Learn More
AdipoGen anti-Polyglutamylation Modification, mAb (GT335) (Biotin)
SDP
Supplier:  AdipoGen AG20B0020BC100
Encompass_Preferred
anti-Polyglutamylation Modification, mAb (GT335) (Biotin), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes the posttranslational modification (poly)glutamylation on proteins. Reacts with polyglutamylated alpha- and beta-tubulin. Isotype: Mouse IgG1kappa. Clone: GT335. Applications: ICC, IP, WB. Liquid. In PBS containing  0.02% sodium azide.
Catalog No. 50-114-5683