Learn More
Medchemexpress LLC Polyglycerol-polyricinoleate | 29894-35-7 | 50 G
SDP
Catalog No. 5000404795
Encompass_Preferred
Polyglycerol-polyricinoleate (PGPR) is an emulsifier commonly used in the food industry. It can improve food texture and stability. This product is for research use only and not sold to patients.
  • Emulsifier commonly used in the food industry
  • Can improve food texture and stability
Catalog No. 50-004-04795 Supplier Medchemexpress LLC Supplier No. HY2935050G
Add to Cart
Add to Cart
Polyglycerol-polyricinoleate (PGPR) is an emulsifier commonly used in the food industry. It can improve food texture and stability. This product is for research use only and not sold to patients.
  • Emulsifier commonly used in the food industry
  • Can improve food texture and stability