Learn More
Cayman Chemical PolygodIal pyrIdazIn 2.5mg
SDP
Catalog No. 502989190
Encompass_Preferred
A derivative of polygodial; inhibits the growth of MCF-7 and Hs 683 cells (GI50s = 72 and 93 μM, respectively)
Catalog No. 50-298-9190 Supplier Cayman Chemical Supplier No. 299002.5 MG
A derivative of polygodial; inhibits the growth of MCF-7 and Hs 683 cells (GI50s = 72 and 93 μM, respectively)