Learn More
Macherey-Nagel TLC polyerster sheets POLYGRAM SIL G UV254, 4x8 cm
Catalog No. NC9578724
Encompass_Preferred
POLYGRAM sheets SIL G/UV254 - size: 4 x 8 cm - pack of 50
Catalog No. NC9578724 Supplier Macherey-Nagel Supplier No. 805021
POLYGRAM sheets SIL G/UV254 - size: 4 x 8 cm - pack of 50