Learn More
Polylc Inc POLYHYDROXYETHYL A COLUMN 5UM
SDP
Catalog No. NC0427094
Encompass
50 x 2.1mm, PolyHYDROXYETHYLA™ columns (5-µm, 60-Å): This neutral, polar material was developed specifically for Hydrophilic Interaction Chromatography (HILIC).
Catalog No. NC0427094 Supplier Polylc Inc Supplier No. 052HY05006
50 x 2.1mm, PolyHYDROXYETHYLA™ columns (5-µm, 60-Å): This neutral, polar material was developed specifically for Hydrophilic Interaction Chromatography (HILIC).