Learn More
MSC POLYIMIDE FILM ELEC TAPE
Catalog No. 5000795939
Encompass
This polyimide film tape with silicone adhesive is designed for PCB solder masking and other high-temperature applications. It is amber colored and 2.7 mil (69 microns) thick.
  • Polyimide film does not soften at elevated temperatures, providing an excellent release surface.
  • Dimensionally stable at high temperatures, reducing rework and enabling high productivity.
  • Flame retardant, chemical, and radiation resistant, protecting surfaces and reducing replacement costs.
  • Silicone adhesive's high temperature performance helps reduce adhesive transfer, eliminating cleaning.
  • Polyethylene tape core instead of cardboard.
Catalog No. 50-007-95939 Supplier MSC Supplier No. 20717823
This polyimide film tape with silicone adhesive is designed for PCB solder masking and other high-temperature applications. It is amber colored and 2.7 mil (69 microns) thick.
  • Polyimide film does not soften at elevated temperatures, providing an excellent release surface.
  • Dimensionally stable at high temperatures, reducing rework and enabling high productivity.
  • Flame retardant, chemical, and radiation resistant, protecting surfaces and reducing replacement costs.
  • Silicone adhesive's high temperature performance helps reduce adhesive transfer, eliminating cleaning.
  • Polyethylene tape core instead of cardboard.