Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Polyimide, 0.0360"" ID x 0.0400"" OD, 12""L; 5/PK
Catalog No. NC9950152
Encompass
The tubing is ideal for use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers. It offers excellent chemical, heat, radiation, and abrasion resistance and high dielectric strength. Resin meets USP Class VI requirements and NEMA standards. The tube has a temperature range of -418 to 896¿F (¿250 to 480¿C). Can be sterilized with autoclave, ethylene oxide or gamma irradiation.
Catalog No. NC9950152 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9582008
The tubing is ideal for use as an electrical and thermal insulator, for fluid and gas delivery, or as a tough sheath for glass fibers. It offers excellent chemical, heat, radiation, and abrasion resistance and high dielectric strength. Resin meets USP Class VI requirements and NEMA standards. The tube has a temperature range of -418 to 896¿F (¿250 to 480¿C). Can be sterilized with autoclave, ethylene oxide or gamma irradiation.