Learn More
Medchemexpress LLC HY-135748 10mg , Polyinosinic-polycytidylic acid (sodium) CAS:42424-50-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13574810MG
Encompass_Preferred
HY-135748 10mg Polyinosinic-polycytidylic acid (sodium) CAS: 42424-50-0 Polyinosinic-polycytidylic acid sodium (Poly(I:C) sodium) is a synthetic analog of double-stranded RNA and an agonist of toll-like receptor 3 (TLR3) and retinoic acid inducible gene I (RIG-I)-like receptors (RIG-I and MDA5). Polyinosinic-polycytidylic acid sodium can be used as a vaccine adjuvant to enhance innate and adaptive immune responses, and to alter the tumor microenvironment. Polyinosinic-polycytidylic acid sodium can directly trigger cancer cells to undergo apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1399