Learn More
TARGETMOL CHEMICALS INC POLYINOSINIC-POLYCYTIDYL 10MG
SDP
Catalog No. 502577422
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Polyinosinic-polycytidylic acid sodium (Poly(IC) sodium) is a synthetic analog of double-stranded RNA and an agonist of toll-like receptor 3 (TLR3) and retinoic acid inducible gene I (RIG-I)-like receptors (RIG-I and MDA5). purity: 98%
Catalog No. 50-257-7422 Supplier TARGETMOL CHEMICALS INC Supplier No. T1251610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Polyinosinic-polycytidylic acid sodium (Poly(IC) sodium) is a synthetic analog of double-stranded RNA and an agonist of toll-like receptor 3 (TLR3) and retinoic acid inducible gene I (RIG-I)-like receptors (RIG-I and MDA5). purity: 98%