Learn More
TARGETMOL CHEMICALS INC POLYINOSINIC-POLYCYTIDYL 25MG
SDP
Catalog No. 502580892
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Polyinosinic-polycytidylic acid (Poly(IC)) is a synthetic double-stranded RNA (dsRNA) analog and is an immunostimulant that acts as the most potent interferon (IFN) inducer. It also is a Toll-like receptor- 3 (TLR3) agonist. purity: 98%
Catalog No. 50-258-0892 Supplier TARGETMOL CHEMICALS INC Supplier No. T2317125MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Polyinosinic-polycytidylic acid (Poly(IC)) is a synthetic double-stranded RNA (dsRNA) analog and is an immunostimulant that acts as the most potent interferon (IFN) inducer. It also is a Toll-like receptor- 3 (TLR3) agonist. purity: 98%