Learn More
Medchemexpress LLC Polyinosinic-polycytidylic acid : kanamycin | 00-00-0 | 51.0% | Mixture of polyinosinic-polycytidylic acid and kanamycin (kanamycin formula C18H36N4O11) | 5 MG
SDP
Catalog No. 5000240611
Encompass_Preferred
Poly (I:C):Kanamycin (1:1) is a research reagent composed of polyinosinic-polycytidylic acid (Poly(I:C)), a synthetic analog of double-stranded RNA, mixed 1:1 with kanamycin. The formulation acts as an immunostimulant by engaging innate immune sensors and is stabilized with kanamycin for handling and storage; it is supplied as a powder in multiple pack sizes with manufacturer-specified storage conditions.
  • Synthetic analog of double-stranded RNA for immune activation.
  • Agonist of Toll-like receptor 3 (TLR3) and RIG-I-like receptors.
  • Used as a vaccine adjuvant and immune stimulant in research.
  • Kanamycin stabilizes the Poly(I:C) to improve handling and shelf life.
  • Available in multiple pack sizes with defined storage recommendations.
  • Powder form stable at low temperature; solvent stability varies by temperature.
Catalog No. 50-002-40611 Supplier Medchemexpress LLC Supplier No. HY107202A5MG
Poly (I:C):Kanamycin (1:1) is a research reagent composed of polyinosinic-polycytidylic acid (Poly(I:C)), a synthetic analog of double-stranded RNA, mixed 1:1 with kanamycin. The formulation acts as an immunostimulant by engaging innate immune sensors and is stabilized with kanamycin for handling and storage; it is supplied as a powder in multiple pack sizes with manufacturer-specified storage conditions.
  • Synthetic analog of double-stranded RNA for immune activation.
  • Agonist of Toll-like receptor 3 (TLR3) and RIG-I-like receptors.
  • Used as a vaccine adjuvant and immune stimulant in research.
  • Kanamycin stabilizes the Poly(I:C) to improve handling and shelf life.
  • Available in multiple pack sizes with defined storage recommendations.
  • Powder form stable at low temperature; solvent stability varies by temperature.