Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Polyisoprene Standards Kit, 10 Vials, 0.5 g, Calibration

Catalog No. 50785371 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Polyisoprene Standards Kit offers a unique collection of 10 GPC (Gel Permeation Chromatography) standards for non-aqueous molecular weight determination.

  • Contents: 10 vials, 0.5 g each, with molecular weight standards (800 to 3,000,000).
  • Molecular weight (Mp) accuracy: < 1,000,000: ± 10&percnt;; > 1,000,000: ± 20&percnt;.
  • Polydispersity (Mw/Mn): Max 1.5.
  • Separation mode: GPC/GFC/SEC.
  • Detection method: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C (Fridge).
  • System type: LC.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Mass: 0.5 g.
  • Type: Calibration.

Catalog No. 50-785-371 Supplier Waters Corp Supplier No. WAT035708
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Polyisoprene Standards Kit offers a unique collection of 10 GPC (Gel Permeation Chromatography) standards for non-aqueous molecular weight determination.

  • Contents: 10 vials, 0.5 g each, with molecular weight standards (800 to 3,000,000).
  • Molecular weight (Mp) accuracy: < 1,000,000: ± 10&percnt;; > 1,000,000: ± 20&percnt;.
  • Polydispersity (Mw/Mn): Max 1.5.
  • Separation mode: GPC/GFC/SEC.
  • Detection method: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C (Fridge).
  • System type: LC.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Mass: 0.5 g.
  • Type: Calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.