Learn More
MSC PIP PolyKor Cut-Resistant Gloves, 13 ga, Small, 6/Pk.
Catalog No. 5000686106
Encompass
These cut, puncture, and abrasion-resistant gloves feature a lightweight PolyKor® blended shell designed to offer excellent dexterity and tactile sensitivity. The seamless construction provides enhanced comfort and breathability for extended wear in various industrial applications.
  • Seamless construction for increased comfort and breathability
  • Polyurethane coated palm and fingers for tactile grip in dry and slightly oily conditions
  • Knit wrist cuff helps prevent dirt and debris from entering the glove
  • Touchscreen compatible design
  • Silicone free and launderable
  • ANSI/ISEA cut level A4 protection
  • Color coded hems for easy size identification
Catalog No. 50-006-86106 Supplier MSC Supplier No. 53877197
These cut, puncture, and abrasion-resistant gloves feature a lightweight PolyKor® blended shell designed to offer excellent dexterity and tactile sensitivity. The seamless construction provides enhanced comfort and breathability for extended wear in various industrial applications.
  • Seamless construction for increased comfort and breathability
  • Polyurethane coated palm and fingers for tactile grip in dry and slightly oily conditions
  • Knit wrist cuff helps prevent dirt and debris from entering the glove
  • Touchscreen compatible design
  • Silicone free and launderable
  • ANSI/ISEA cut level A4 protection
  • Color coded hems for easy size identification