Learn More
AdipoGen PolyL-lactide 10 dl g
SDP
Supplier:  AdipoGen CDXP0013G010
Encompass_Preferred
Chemical. CAS 26161-42-2. Formula C3H4O2n. MW 59,000-101,000. Synthetic. Polylactides PLA used in material sciences. Biodegradable Polymer. Lactide and Glycolide Polymers.
Catalog No. 50-256-0286