Learn More
Medchemexpress LLC Polylactic acid | 26100-51-6 | MFCD00081867 | ≥98.0% | 60,000 (average) | (C3H6O3)x | 1 G
SDP
Catalog No. 5000280642
Encompass_Preferred
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester commonly used as a structural polymer, scaffold material, and drug delivery carrier in biomedical research and materials science. It is supplied for laboratory use with high purity and is suitable for formulation, processing, and degradation studies.
  • High purity (≥98.0%) suitable for research applications.
  • Biodegradable and biocompatible polymer for implants and scaffolds.
  • Average molecular weight approximately 60,000 for mechanical strength.
  • Processable in common organic solvents for fabrication and formulation.
  • Available in multiple laboratory pack sizes for experimentation.
Catalog No. 50-002-80642 Supplier Medchemexpress LLC Supplier No. HYW2503131G
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester commonly used as a structural polymer, scaffold material, and drug delivery carrier in biomedical research and materials science. It is supplied for laboratory use with high purity and is suitable for formulation, processing, and degradation studies.
  • High purity (≥98.0%) suitable for research applications.
  • Biodegradable and biocompatible polymer for implants and scaffolds.
  • Average molecular weight approximately 60,000 for mechanical strength.
  • Processable in common organic solvents for fabrication and formulation.
  • Available in multiple laboratory pack sizes for experimentation.