Learn More
Medchemexpress LLC Polylactic acid | 26100-51-6 | MFCD00081867 | ≥98.0% | ≈60,000 g/mol | (C3H6O3)x | 500 MG
SDP
Catalog No. 5000280616
Encompass_Preferred
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester used as a matrix for controlled drug release and as a structural material in tissue engineering. It hydrolyzes to lactic acid, allowing tunable degradation and mechanical properties for formulations and scaffolds used in drug delivery, orthopedic fixation, and 3D-printed bone regeneration.
  • Biodegradable and biocompatible polymer.
  • Enables controlled release via hydrolytic degradation.
  • Tunable degradation rate and mechanical properties.
  • Compatible with composite formulations and 3D printing.
  • Solid form suitable for scaffold and implant fabrication.
  • High purity (≥98.0%).
Catalog No. 50-002-80616 Supplier Medchemexpress LLC Supplier No. HYW250313500MG
Polylactic acid (PLA) is a biodegradable, biocompatible aliphatic polyester used as a matrix for controlled drug release and as a structural material in tissue engineering. It hydrolyzes to lactic acid, allowing tunable degradation and mechanical properties for formulations and scaffolds used in drug delivery, orthopedic fixation, and 3D-printed bone regeneration.
  • Biodegradable and biocompatible polymer.
  • Enables controlled release via hydrolytic degradation.
  • Tunable degradation rate and mechanical properties.
  • Compatible with composite formulations and 3D printing.
  • Solid form suitable for scaffold and implant fabrication.
  • High purity (≥98.0%).