Learn More
Medchemexpress LLC Polylactic acid | 26100-51-6 | MFCD00081867 | ≥98.0% | (C3H6O3)x | 10 G
SDP
Catalog No. 5000280507
Encompass_Preferred
Polylactic acid (PLA) is a biodegradable, biocompatible polyester used in research for drug delivery, tissue engineering, and as a structural polymer. This 10 g laboratory reagent is supplied at high purity for formulation and material science applications.
  • High purity (≥98.0%) suitable for research
  • Average molecular weight ≈60,000 g/mol for typical polymer behavior
  • White to off-white solid convenient for handling
  • Processable for formulation, scaffold fabrication, and 3D printing studies
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month)
  • CAS number 26100-51-6 for unambiguous identification
Catalog No. 50-002-80507 Supplier Medchemexpress LLC Supplier No. HYW25031310G
Polylactic acid (PLA) is a biodegradable, biocompatible polyester used in research for drug delivery, tissue engineering, and as a structural polymer. This 10 g laboratory reagent is supplied at high purity for formulation and material science applications.
  • High purity (≥98.0%) suitable for research
  • Average molecular weight ≈60,000 g/mol for typical polymer behavior
  • White to off-white solid convenient for handling
  • Processable for formulation, scaffold fabrication, and 3D printing studies
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month)
  • CAS number 26100-51-6 for unambiguous identification