Description
- Test uses monoclonal antibodies to detect presence of bladder tumor associated antigen in urine
- Detects early stage and grade cancers that cytology often misses
- Returns fast, objective result, a line in patient window indicates positive result
- Requires only one voided urine sample, with no sample preparation
- Adjunct test to cystoscopy
Specifications
Specifications
| Type | BTA stat™ Test Kit |
| Includes | 10 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 2 Instructions for Home Use |
| Quantity | 10 Tests |