Learn More
Polymedco™ Polymedco BTA stat™ Test Kit
SDP
Catalog No. 21000132 Shop All Polymedco Products
Change view
Click to view available options
Includes:
10 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 2 Instructions for Home Use
30 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 5 Instructions for Home Use
Quantity:
10 Tests
30 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
21-000-132 30 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 5 Instructions for Home Use 30 Tests
21-000-131 10 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 2 Instructions for Home Use 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-000-132 Supplier Polymedco™ Supplier No. 661230
Add to Cart
Add to Cart
This item is not returnable. View return policy
For detection of recurrent bladder cancer in five minutes
  •  Test uses monoclonal antibodies to detect presence of bladder tumor associated antigen in urine
  • Detects early stage and grade cancers that cytology often misses
  • Returns fast, objective result, a line in patient window indicates positive result
  • Requires only one voided urine sample, with no sample preparation
  • Adjunct test to cystoscopy
Specifications
Type BTA stat™ Test Kit
Includes 30 Foil Packages. Each package contains - 1 BTA stat device, 1 Disposable dropper, 1 Disposable desiccant pouch, 1 Package Insert, 5 Instructions for Home Use
Quantity 30 Tests