Learn More
Polymedco™ Poly stat™ hCG Pregnancy Test
SDP
Catalog No. 21000127
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
21-000-127 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 21-000-127 Supplier Polymedco™ Supplier No. HCG25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Chromatographic immunoassay for rapid qualitative determination of hCG in serum and urine
Certifications
CLIA Complexity: Waived (Urine), Moderate (Serum)
Specifications
Clia Complexity Waived, Moderate
Detectable Analytes hCG
Sample Type Serum, Urine
Type hCG Test Kit
Quantity 25 Tests