Learn More
Polymedco™ Poly stat™ H. pylori test
SDP
Catalog No. 21000126 Shop All Polymedco Products
Change view
Click to view available options
Quantity:
20 Tests
Catalog No. Quantity
21-000-126 20 Tests
1 options
This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.
Catalog No. 21-000-126 Supplier Polymedco™ Supplier No. HP20
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.
For rapid qualitative detection of IgG antibodies specific to Helicobacter pylori in human serum, plasma and whole blood specimens
Certifications
CLIA Complexity: Waived (Whole blood), Moderate (Serum or plasma)
Specifications
Clia Complexity Waived, Moderate
Detectable Analytes H. pylori IgG
Type H. pylori Test
Quantity 20 Tests