Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Poly stat™ Mono test
SDP

Catalog No. 21000128 Shop All Polymedco Products
Change view
Click to view available options
Quantity:
20 Cassettes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-000-128 20 Cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.
Catalog No. 21-000-128 Supplier Polymedco™ Supplier No. MN20
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.

Rapid test for visual qualitative detection of heterophile antibodies specific to Infectious Mononucleosis (IM) in human serum, plasma or whole blood

Certifications

CLIA Complexity: Waived (Whole blood), Moderate (Serum or plasma)

Specifications

Clia Complexity Waived, Moderate
Detectable Analytes Mononucleosis Heterophil Antibodies
Format Cassette
Type Infectious Mononucleosis Test
Quantity 20 Cassettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.