Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Poly stat™ Strep A Test
SDP

Catalog No. 21000129 Shop All Polymedco Products
Change view
Click to view available options
Format:
Flip
Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
21-000-129 Strip
21-000-130 Flip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.
Catalog No. 21-000-129 Supplier Polymedco™ Supplier No. STS25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.

Lateral flow, rapid one-step test for qualitative detection of Group A Streptococcal antigen

  • Direct detection from throat swabs
  • Simple visual result
  • Available in flip and strip formats

Certifications

CLIA Complexity: Waived (Flip format), For waived facilities (Strip format)

Specifications

Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Format Strip
Sample Type Throat Swab
Type Poly Stat Streptococcus A Test
Quantity 25 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.