Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Sed-Chek™ 2 ESR Controls for StaRRsed Analyzers (Select Models)
SDP

Catalog No. 21000121 Shop All Polymedco Products
Change view
Click to view available options
Type:
Abnormal Control
Normal Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
21-000-121 Normal Control
21-000-120 Abnormal Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-000-121 Supplier Polymedco™ Supplier No. ESR2SSN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Whole blood reference control materials designed to monitor ESR procedures

Controls help to monitor technique as well as environmental, physical, and mechanical factors such as room temperature, and tube position, vibration, or instrument problems. With an advanced formula, these controls are comprehensive and assayed for both automated and manual methods.

  • 18 month closed-vial stability from date of manufacture
  • 31 day open-vial stability
  • Easy-to-mix formula
  • Available in 6 x 5mL vials

Specifications

Cap Type Automatic self-zeroing
Compatible Methods Both automated and manual ESR methods
Content And Storage 20°C to 22°C
For Use With (Equipment) StaRRsed analyzer
Type Normal Control
Intended Use Whole Blood reference control material designed to monitor ESR procedures
Packaging Vial
Quantity 6 x 5 mL
Stability 31 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.