Learn More
Fisherfinest™ Sedimat™ 15 Plus Sedrate System
SDP
Catalog No. 21000142 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Sedimat 15 Plus Sedrate System
Sedmat 15 Plus Base
For Use With (Equipment):
LIS compatible units
Non-LIS compatible units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
21-000-142 Sedimat 15 Plus Sedrate System -
21-000-143 Sedmat 15 Plus Base Non-LIS compatible units
21-000-149 Sedmat 15 Plus Base LIS compatible units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 21-000-142
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fast, in-office results for one to eight specimens simultaneously.
Automated sedrate system provides accurate ESR results in only 15 minutes.
  • Random access capability
  • Faster throughput; processes 32 patient results per hour
  • Eight-position reader
  • Small footprint suitable for tight benchtops
  • Bar code reader available(21-000-145 )
  • LIS compatible for data transfer
Specifications
Type Sedimat 15 Plus Sedrate System
For Use With (Application) Fast, in-office results for one to eight specimens simultaneously
Sample Size 0.8 mL
Throughput 32 Samples/Hour
Frequency 60 Hz
Voltage 120 V