Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ Venoplast™ Vacuum and Nonvacuum ESR Tubes
SDP

Catalog No. 21000108 Shop All Polymedco Products
Change view
Click to view available options
Product Type:
ESR Non-vacuum Tube no Diluent
ESR Non-vacuum Tube w/ Diluent
ESR Vacuum Tube with Diluent
Quantity:
1000 Box
200 Box
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
21-000-108 ESR Vacuum Tube with Diluent 200 Box
21-000-109 ESR Vacuum Tube with Diluent 1000 Box
21-000-111 ESR Non-vacuum Tube w/ Diluent 1000 Box
21-000-110 ESR Non-vacuum Tube w/ Diluent 200 Box
21-000-112 ESR Non-vacuum Tube no Diluent 200 Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-000-108 Supplier Polymedco™ Supplier No. 265277
Add to Cart
Add to Cart

For use with Polymedco Sediten 10-Minute ESR Reader

Available vacuum or nonvacuum and with or without diluent.

Specifications

Product Type ESR Vacuum Tube with Diluent
For Use With (Equipment) Polymedco Sediten 10-Minute ESR Reader
Quantity 200 Box
Includes Diluent
Certifications/Compliance US Federal and state regulations
Media Type 3.8% Sodium Citrate
pH 5.65
Format Clear Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.