Learn More
Anton Paar Polymer melt rheometer for QC and routine measurementsSmallest footprint and most sustainable operationHighest-performing electrical oven on the market (-150 °C to +400 °C)Minimum training requirements
Catalog No. 5000580555
Encompass_Preferred
MCR 73 SmartMelt SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: n/aMin. torque (rotation): 0.1 μNmMin. torque (oscillation): 0.1 μNmMax. torque: 160 mNmMinimum angular deflection (set value): 1 μradMaximum angular deflection (set value): ∞ μradMaximum speed: 2,500 rpmMinimum angular frequency: 2 x 10-5 HzMaximum angular frequency: 100 HzNormal force measurement design: n/aNormal force range: n/aTemperature deviceTemperature oven design: Electrical temperature ovenRecommended measuring geometry: Plate-plate, disposablesTemperature range: -150 °C to +400 °C Maximum heating rate: 50 °C/minMaximum cooling rate: Up to 100 °C/min2)Fully automatic temperature calibration: yes
Catalog No. 50-005-80555 Supplier Anton Paar Supplier No. 385254
Please contact your sales representative for a quote or further ordering instructions.
MCR 73 SmartMelt SpecificationsBearing design: Air, fine-pored carbonMotor design: Electronically commutated (EC) permanent magnet synchronous motorDisplacement transducer: High-resolution optical encoderNormal force measurement design: n/aMin. torque (rotation): 0.1 μNmMin. torque (oscillation): 0.1 μNmMax. torque: 160 mNmMinimum angular deflection (set value): 1 μradMaximum angular deflection (set value): ∞ μradMaximum speed: 2,500 rpmMinimum angular frequency: 2 x 10-5 HzMaximum angular frequency: 100 HzNormal force measurement design: n/aNormal force range: n/aTemperature deviceTemperature oven design: Electrical temperature ovenRecommended measuring geometry: Plate-plate, disposablesTemperature range: -150 °C to +400 °C Maximum heating rate: 50 °C/minMaximum cooling rate: Up to 100 °C/min2)Fully automatic temperature calibration: yes