Learn More
PTS Diagnostics A1CNow+™ Professional Test System
Catalog No. 21124415
Change view
Click to view available options
Quantity:
10-Count Test Kit
20-Count Test Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-124-415 10-Count Test Kit
21-124-414 20-Count Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-124-415 Supplier PTS Diagnostics Supplier No. 3024
Add to Cart
Add to Cart
This item is not returnable. View return policy
A fast and easy way of obtaining accurate A1C results
  • Accurate A1C results in as little as 5 minutes.
  • For professional use to monitor glycemic control in people with diabetes.
  • Uses a small 5 μL fingerstick blood sample.
  • Can be stored at room temperature for 4 months.
  • Provides quantitative % A1C results that reflect average blood sugar control over the past 2-3 months.
  • Dispose of analyzer when all tests in the box have been used.
  • Box includes one A1CNow+ analyzer, 20 test cartridges, and 20 blood shaker/collectors.
Certifications
National Glycohemoglobin Standardization Program (NGSP), IFCC
Specifications
Type Hemoglobin Analyzer
Sample Type Blood
Quantity 10-Count Test Kit