Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTS Diagnostics Unistik™ 3 Extra Single Use Safety Lancets

Catalog No. 21124407
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
21-124-407 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-407 Supplier PTS Diagnostics Supplier No. 1701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Simple capillary blood sampling with single-use Unistik 3 lancets.Comfort Zone Technology™

  • Reduce risk of infection.
  • Extra model - 21 gauge, 2.0 mm depth with 75 to 125 μL blood volume range
  • Simple - one-click operation
  • Safe - needles retracts immediately after use
  • Sterile - single use, sterile device
  • Accurate - targeted positioning
  • Lockout indicator - prevents re-use

Specifications

Depth (Metric) Needle 2.0 mm
Flow Type High
Needle Gauge 21
Packaging Quantity 100/Box
Sterility Sterile
Color Orange
Disposable Single-use
For Use With (Application) For diagnostic tests that require more than a standard drop of blood
Product Type Lancet
For Use With (Equipment) CardioChek analyzers and PTS panels test strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.