Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Polymers - Making Fluorescent Worms
GREENER_CHOICESCI_ED

Catalog No. S20641 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S20641 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including D&C Red No. 19, which is known to the State of California to cause cancer.
Catalog No. S20641 Supplier Innovating Science™ Supplier No. IS7045
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including D&C Red No. 19, which is known to the State of California to cause cancer.

Requires

3 Containers for calcium chloride solutions (such as beakers, glass jars, or plastictrays) Black light

Allow students to learn about polymers and cross-linking using this fun and colorful activity.

  • Students add a solution of sodium alginate to three calcium chloride solutions, each containing a different fluorescent dye
  • After a few minutes the liquid sodium alginate solution forms into gel-like polymer worms
  • Upon completion of the activity, the worms may be placed under a black light and the students will observe their worms glow red, blue, and green
  • Contains enough material to make several dozen polymer worms

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes 6 g Sodium Alginate, 1 L Calcium Chloride/0.1% Fluorescein, 0.1M, 1 L Calcium Chloride/0.04% Rhodamine B, 0.1M, 1 L Calcium Chloride/0.5% Quinine Sulfate, 0.1M, 3 Empty bottles with flip-top caps, 6 Forceps
Quantity 1
Dimensions (L x W x H) 7 x 7.25 x 12.375 in.
Class Size 1 Class
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.