Learn More
Invivogen POLYMIXIN B - 100 MG
SDP
Catalog No. NC9299687
Encompass
Polymixin B; Inhibitor of LPS-induced TLR4 activation; White powder; 100mg; 1 385; C 55 H 96 N 16 O 13; 2H2SO4; pH: 5 to 7;5; 1405-20-5; Store in a dry place at ambient pressure
Catalog No. NC9299687 Supplier Invivogen Supplier No. TLRLPMB
Polymixin B; Inhibitor of LPS-induced TLR4 activation; White powder; 100mg; 1 385; C 55 H 96 N 16 O 13; 2H2SO4; pH: 5 to 7;5; 1405-20-5; Store in a dry place at ambient pressure