Learn More
Research Products International Corp POLYMIXIN B SULFATE, 1GM
SDP
Supplier:  Research Products International Corp P401601.0
Encompass_Preferred
Polymyxin B Sulfate is an antibiotic that belongs to the polymyxin class of antibiotics and is primarily effective against Gram-negative bacteria. 
Polymyxin B is derived from Bacillus polymyxa and acts by disrupting the integrity of the bacterial cell membrane. 
In the field of microbiology and cell culture, Polymyxin B Sulfate is commonly used to prevent bacterial contamination. It is particularly effective against Gram-negative bacteria, including multidrug-resistant strains such as Pseudomonas aeruginosa. Researchers add Polymyxin B to growth media or solutions to inhibit the growth of these bacteria and maintain the purity of cultures. 
Overall, Polymyxin B Sulfate is a valuable antibiotic in the life science marketplace, primarily used in microbiology, cell culture, and molecular biology research.
Catalog No. 50-213-625