Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Polymixin B sulfate, 50 mg/ml in distilled water, sterile-filtered

Catalog No. AAJ67163XF
Change view
Click to view available options
Quantity:
1mL
5 x 1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67163XF 1mL
AAJ671638EQ 5 x 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67163XF Supplier Thermo Scientific Chemicals Supplier No. J67163XF
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-20-5

Specifications

Content And Storage Store at -20°C
Molecular Formula C48H84N16O17S
Format Liquid
MDL Number MFCD00131991
Molecular Weight (g/mol) 1301.571, 1189.36
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O
IUPAC Name 4-amino-N-{1-[(3-amino-1-{[6,9,18-tris(2-aminoethyl)-15-benzyl-3-(2-hydroxyethyl)-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclotricosan-21-yl]carbamoyl}propyl)carbamoyl]-3-hydroxypropyl}-2-formamidobutanamide; sulfuric acid
Product Type Polymixin B sulfate
Quantity 1mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.