Learn More
Cosmo Bio Usa Inc Polymorphprep™ 250ml
SDP
Catalog No. NC0863559
Encompass
Polymorphprep™ is a ready-made, sterile, and endotoxin tested solution used to isolate polymorphonuclear leukocytes. Polymorphprep™ specifications:Sodium diatrizoate: 13.8% (w/v) Dextran 500: 8.0% (w/v) Density: 1.113 ± 0.001 µg/ml; Osmolality: 445 ± 15 mOsm; Endotoxins: < 1.0 EU/ml ; Polymorphprep™ is the ideal medium for: polymorphonuclear leukocytes
Catalog No. NC0863559 Supplier Cosmo Bio Usa Inc Supplier No. AXS1114683
Polymorphprep™ is a ready-made, sterile, and endotoxin tested solution used to isolate polymorphonuclear leukocytes. Polymorphprep™ specifications:Sodium diatrizoate: 13.8% (w/v) Dextran 500: 8.0% (w/v) Density: 1.113 ± 0.001 µg/ml; Osmolality: 445 ± 15 mOsm; Endotoxins: < 1.0 EU/ml ; Polymorphprep™ is the ideal medium for: polymorphonuclear leukocytes