Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Cold Mount Polymount Kit Polyester Resin
SDP

Supplier:  Electron Microscopy Sciences 1270

Encompass_Preferred

500 ml liquid resin; 15 ml hardening liquid. Use for serial mounting of specimens not sensitive to heat. Polyester belongs to the catalyzed system, like acrylics. Curing times are relatively short and the hardened specimen is duroplastic.
Color: Transparent
Curing time: 45 minutes
Use: Serial mounting of specimens not sensitive to heat
Shrinkage: 4

Catalog No. 50-980-336


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.