Learn More
Polysciences, Inc. Poly-Mount Xylene, 120 mL
SDP
Catalog No. NC0825496
Encompass_Preferred
Xylene based routine coverslipping medium for histology and cytology slides. This acrylic medium provides superior optical clarity. Poly-Mount Xylene is also recommended for automated coverslipping instruments that use glass coverslips. It contains an anti-fade agent to prevent fading and yellowing with long term storage. Dries quickly for viewing, storage, and filing.
Catalog No. NC0825496 Supplier Polysciences, Inc. Supplier No. 24176120
Xylene based routine coverslipping medium for histology and cytology slides. This acrylic medium provides superior optical clarity. Poly-Mount Xylene is also recommended for automated coverslipping instruments that use glass coverslips. It contains an anti-fade agent to prevent fading and yellowing with long term storage. Dries quickly for viewing, storage, and filing.