Learn More
Medchemexpress LLC HY-106783A 100mg , Polymyxin B nonapeptide (TFA) CAS:2220175-42-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106783A100MG
Encompass_Preferred
HY-106783A 100mg Polymyxin B nonapeptide (TFA) CAS: 2220175-42-6 Polymyxin B nonapeptide TFA is a cyclic peptide obtained from Polymyxin B by proteolytic removal of its terminal amino acyl residue. Polymyxin B nonapeptide TFA is less toxic, lacks bactericidal activity, and retains its ability to render gram-negative bacteria susceptible to several antibiotics by permeabilizing their outer membranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9623