Learn More
Medchemexpress LLC Polymyxin B Sulfate | 1405-20-5 | 96.9% | C111H196N32O30S | 1 G
SDP
Catalog No. 5000432265
Encompass_Preferred
Polymyxin B Sulfate is a potent antibacterial agent and a relatively toxic antibiotic. It also functions as an antiendotoxin agent, exhibiting endotoxin-neutralizing properties that can be utilized in adjunctive research for gram-negative sepsis. It demonstrates antibacterial activities both in vitro and in vivo.
  • Potent antibacterial agent
  • Exhibits antiendotoxin activity
  • Useful in adjunctive research for gram-negative sepsis
  • Demonstrates antibacterial activity in vitro
  • Demonstrates antibacterial activity in vivo
  • Targets bacterial pathogens
  • Part of anti-infection pathways
  • Suitable for mammalian cell culture research
Catalog No. 50-004-32265 Supplier Medchemexpress LLC Supplier No. HYA02481G
Polymyxin B Sulfate is a potent antibacterial agent and a relatively toxic antibiotic. It also functions as an antiendotoxin agent, exhibiting endotoxin-neutralizing properties that can be utilized in adjunctive research for gram-negative sepsis. It demonstrates antibacterial activities both in vitro and in vivo.
  • Potent antibacterial agent
  • Exhibits antiendotoxin activity
  • Useful in adjunctive research for gram-negative sepsis
  • Demonstrates antibacterial activity in vitro
  • Demonstrates antibacterial activity in vivo
  • Targets bacterial pathogens
  • Part of anti-infection pathways
  • Suitable for mammalian cell culture research