Learn More
Enzo Life Sciences Polymyxin B . sulfate (5g). CAS: 1405-20-5
SDP
Catalog No. 502008926
Encompass_Preferred
Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase. Purity: ≥80% (HPLC) (sum of polymyxins B1, B2, B3, B1-I). Appearance: White to off-white powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8926 Supplier Enzo Life Sciences Supplier No. ALX380040G005
Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase. Purity: ≥80% (HPLC) (sum of polymyxins B1, B2, B3, B1-I). Appearance: White to off-white powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.