Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymyxin B Sulfate, G-Biosciences
SCI_ED

Catalog No. S157219
Change view
Click to view available options
Quantity:
10 MU
100 MU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157219 10 MU
S139788 100 MU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157219 Supplier G-Biosciences Supplier No. RC1091
Add to Cart
Add to Cart

Polymyxin B sulfate is a polypeptide antibiotic mixture of B1, B2, and B3 polymyxins. Each of these fractions vary by a fatty acid group and exhibit marginal MIC differences when compared.

Polymyxin B is most effective against gram-negative bacteria. It has been shown to work against Pseudomonas aeruginosa, Pseudomonas viridiflava, and Erwinia carotovora. Polymyxin B is freely soluble in aqueous solution.

Chemical Identifiers

Molecular Weight (g/mol) 1385.61 g/mol

Specifications

Quantity 10 MU
Percent Purity >80%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.