Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymyxin B Sulfate MP Biomedicals

Catalog No. ICN10056510 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mU
25 mU
5 mU
50 mU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10056510 10 mU
ICN10056505 5 mU
ICN10056525 25 mU
ICN10056550 50 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10056510 Supplier MP Biomedicals, Inc Supplier No. 0210056510
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1405-20-5
Molecular Formula C48H84N16O17S
Molecular Weight (g/mol) 1189.36
MDL Number MFCD00131991
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
Synonym Aerosporin
PubChem CID 133109994
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O

Specifications

Type Polymixin B Sulfate
Physical Form Powder
Color White
Quantity 10 mU
Formula Weight 1385.61
Chemical Name or Material Polymyxin B Sulfate, Aerosporin Activity: >6, 000 u/mg
Recommended Storage Store at 2° to 8°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.