Learn More
Sigma Aldrich Fine Chemicals Biosciences Polymyxin B sulfate salt p
Catalog No. 502897387
Encompass
Polymyxin B is a member of the polymyxin group of antibiotics obtained from different strains of Bacillus polymyxa and related species. It contains a cyclic heptapeptide unit and a tripeptide side chain which is N-acylated by a fatty acyl residue. Polymyxin B sulfate salt is generally used for the treatment of Gram-negative bacterial infections.Chemical structure peptide
Catalog No. 50-289-7387 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P49321MU
Polymyxin B is a member of the polymyxin group of antibiotics obtained from different strains of Bacillus polymyxa and related species. It contains a cyclic heptapeptide unit and a tripeptide side chain which is N-acylated by a fatty acyl residue. Polymyxin B sulfate salt is generally used for the treatment of Gram-negative bacterial infections.Chemical structure peptide