Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymyxin B Sulfate USP Micronized, Medisca™
GREENER_CHOICESDP

Catalog No. 18048082 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-082 100 μg
18-048-043 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-048-082 Supplier Medisca Supplier No. 004503
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1405-20-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Color White to Off-White
Quantity 100 μg
Percent Purity ≥ 6000 Units/mg (Polymyxin B)
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.