Learn More
Medchemexpress LLC Polymyxin B1 | 4135-11-9 | MFCD01699017 | 98.90% | C56H98N16O13 | 1 MG
SDP
Catalog No. 5000432393
Encompass_Preferred
Polymyxin B1 is a potent antimicrobial lipopeptide derived from Bacilus polymyxa. It is the major component in Polymyxin B and functions by inducing lysis of bacterial cells through interaction with their membranes. It holds potential for treating multidrug-resistant Gram-negative bacterial infections.
  • Potent antimicrobial lipopeptide
  • Derived from Bacilus polymyxa
  • Major component in Polymyxin B
  • Induces bacterial cell lysis by interacting with membranes
  • Potential for treating multidrug-resistant gram-negative bacterial infections
  • Binds to negatively charged sites in bacterial cell membranes
  • Disrupts cell wall integrity and increases permeability for water and molecules
Catalog No. 50-004-32393 Supplier Medchemexpress LLC Supplier No. HYA0248A1MG
Polymyxin B1 is a potent antimicrobial lipopeptide derived from Bacilus polymyxa. It is the major component in Polymyxin B and functions by inducing lysis of bacterial cells through interaction with their membranes. It holds potential for treating multidrug-resistant Gram-negative bacterial infections.
  • Potent antimicrobial lipopeptide
  • Derived from Bacilus polymyxa
  • Major component in Polymyxin B
  • Induces bacterial cell lysis by interacting with membranes
  • Potential for treating multidrug-resistant gram-negative bacterial infections
  • Binds to negatively charged sites in bacterial cell membranes
  • Disrupts cell wall integrity and increases permeability for water and molecules