Learn More
Cayman Chemical PolymyxIn B1 IsoleucInsulf
SDP
Catalog No. 502955589
Encompass_Preferred
Differs from polymyxin B1 by having isoleucine rather than leucine as a component of the cyclic peptide; comparable to other B polymyxins in its ability to kill P. aeruginosa, A. baumannii, and K. pneumoniae isolates
Catalog No. 50-295-5589 Supplier Cayman Chemical Supplier No. 14075500 UG
Differs from polymyxin B1 by having isoleucine rather than leucine as a component of the cyclic peptide; comparable to other B polymyxins in its ability to kill P. aeruginosa, A. baumannii, and K. pneumoniae isolates