Learn More
Cayman Chemical Polymyxin B (sulfate), Purity: ≥80% (B Polymyxins), CAS: 1405-20-5, 1 g
SDP
Catalog No. 502953601
Encompass_Preferred
A mixture of at least four closely related components, polymyxin B1 to B4, with polymyxin B1 and B2 being the two major components; demonstrates rapid in vitro bactericidal activity against major multidrug-resistant Gram-negative bacteria, including P. aeruginosa, A. baumannii, and K. pneumonia
Catalog No. 50-295-3601 Supplier Cayman Chemical Supplier No. 141571 G
A mixture of at least four closely related components, polymyxin B1 to B4, with polymyxin B1 and B2 being the two major components; demonstrates rapid in vitro bactericidal activity against major multidrug-resistant Gram-negative bacteria, including P. aeruginosa, A. baumannii, and K. pneumonia