Learn More
Polynuclear Aromatic Hydrocarbons 16 Solution, Analytical Standard, in Acetonitrile, MilliporeSigma™ Supelco™
Catalog No. 111017103 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7103 2 mL
1 options
Catalog No. 11-101-7103 Supplier MilliporeSigma Supelco Supplier No. 369792ML
Specifications
Description 10 μg each/mL acenaphthene, acenaphthylene, anthracene, benzo[a]anthracene, benzo[b]fluoranthene, benzo[k]fluoranthene, benzo[ghi]perylene, benzo[a]pyrene, chrysene, dibenzo[a,h]anthracene, fluoranthene, fluorene, indeno[1,2,3-cd]pyrene, naphthalene, phenanthrene and pyrene
Quantity 2 mL
Grade Analytical Standard