Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polynuclear Aromatic Hydrocarbons, SPEX CertiPrep™
SDP

Catalog No. CLPSB Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPS-B 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPS-B Supplier SPEX CertiPrep Supplier No. CLPSB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Naphthalene91-20-32000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material PAH Analyte Mix
Packaging 2mL amber prescored glass ampule
Name Note EPA Methods 610, 625, SW-846 Methods 8100, 8270
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA Methods 610, 625, 8100, 8270, 8270C
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Methylene Chloride/Benzene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Components at 2000μg/mL in Methylene Chloride:Benzene:
Acenaphthene; Acenaphthylene; Anthracene; Benzo(a)anthracene; Benzo(a)pyrene; Benzo(b)fluoranthene; Benzo(g,h,i)perylene; Benzo(k)fluoranthene; Dibenz(a,h)anthracene; Chrysene; Fluoranthene; Fluorene; Indeno(1,2,3-cd)pyrene; Naphthalene; Phenanthrene; Pyrene
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.