Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyoxyethanyl-Alpha-Tocopheryl Sebacate

Catalog No. 69871710ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-871-710ML 10 mL
69-871-7100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-871-710ML Supplier Sigma Aldrich Supplier No. 69871710ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced to increase the catalytic efficiency and acts as an environmentally benign and sustainable amphiphile. Click here for more information.PTS: Amphiphile for Organic Synthesis in WaterUses:Provides an aqueous micellar environment for transition metal-catalyzed Heck coupling, cross metathesis and ring closing metathesis reactionsSurfactant for catalytic asymmetric Baeyer-Villiger oxidation in water using PtII catalysts and hydrogen peroxideOrally active CoQ10 carrier for neuroprotection after stroke or cardiac arrestProduct of Zymes LLC sold under exclusive license to US and worldwide patents.

Specifications

CAS 263015-34-5
Quantity 10 mL
Synonym PTS
Molecular Weight (g/mol) ∼1,200 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.