Learn More
Medchemexpress LLC Polyoxyethylene (10) lauryl ether | 9002-92-0 | 98.0% | 100 G
SDP
Catalog No. 5000411022
Encompass_Preferred
Polyoxyethylene (10) lauryl ether is a nonionic surfactant. It exhibits unique properties in dissolving liposomes and exerts minimal interference with endotoxin detection. It forms hybrid liposomes with Dimyristoylphosphatidylcholine (DMPC), which can inhibit the proliferation of human promyelocytic leukemia cells without the administration of any drugs.
  • For research use only. We do not sell to patients.
  • Enhanced the anti-tumor activity of DMPC and its hybrid liposomes activated caspase-3/8/9 and Fas, inducing apoptosis in HL-60 cells.
Catalog No. 50-004-11022 Supplier Medchemexpress LLC Supplier No. HYB2106D100G
Polyoxyethylene (10) lauryl ether is a nonionic surfactant. It exhibits unique properties in dissolving liposomes and exerts minimal interference with endotoxin detection. It forms hybrid liposomes with Dimyristoylphosphatidylcholine (DMPC), which can inhibit the proliferation of human promyelocytic leukemia cells without the administration of any drugs.
  • For research use only. We do not sell to patients.
  • Enhanced the anti-tumor activity of DMPC and its hybrid liposomes activated caspase-3/8/9 and Fas, inducing apoptosis in HL-60 cells.